Stampa…

(tert-Butyldimethylsilyl)acetylene ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

B4365-5G B4365-25G
TCIAB4365-5GEA 99 EUR
TCIAB4365-5G TCIAB4365-25G
(tert-Butyldimethylsilyl)acetylene ≥97.0% (by GC)
(tert-Butyldimethylsilyl)acetylene
Formula: C₈H₁₆Si
Peso molecolare: 140,3 g/mol
Punto di ebollizione: 110°C
Flash Pt: 6°C
MDL Number: MFCD00191877
CAS Number: 86318-61-8
UN: 1993
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR