Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
B4360-1G
B4360-5G
TCIAB4360-1GEA
111
EUR
TCIAB4360-1G
TCIAB4360-5G
Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) ≥98.0% (by HPLC)
Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II)
Formula:
C₃₂H₂₈NiO₄S₄ Peso molecolare: 663,51 g/mol |
MDL Number:
MFCD10567145 CAS Number: 38951-97-2 EINECS: 254-212-5 |
Risultati del test di specificazione
Appearance | Black powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Chelometric Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...