(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid
Formula:
C₁₅H₂₁NO₄ Peso molecolare: 279.34 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00076904 CAS Number: 100564-78-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Specific rotation [a]20/D | +5.0 to +7.0 deg (C=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...