5-Bromo-6-chloronicotinic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
B4355-1G
B4355-5G
TCIAB4355-1GEA
27
EUR
TCIAB4355-1G
TCIAB4355-5G
5-Bromo-6-chloronicotinic acid ≥98.0% (by GC, titration analysis)
5-Bromo-6-chloronicotinic acid
Formula:
C₆H₃BrClNO₂ |
MDL Number:
MFCD01927098 CAS Number: 29241-62-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...