Stampa…

9-Benzyl-3,6-dibromocarbazole ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE

B4348-1G B4348-5G
TCIAB4348-1GEA 77 EUR
TCIAB4348-1G TCIAB4348-5G
9-Benzyl-3,6-dibromocarbazole ≥98.0% (by GC, total nitrogen)
9-Benzyl-3,6-dibromocarbazole
Formula: C₁₉H₁₃Br₂N
MDL Number: MFCD00218292
CAS Number: 118599-27-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR