Stampa…

4-Bromodibenzofuran ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Bromodibenzo[b,d]furan

B4345-1G B4345-5G
TCIAB4345-1GEA 53 EUR
TCIAB4345-1G TCIAB4345-5G
4-Bromodibenzofuran ≥98.0% (by GC)
4-Bromodibenzofuran
Formula: C₁₂H₇BrO
Peso molecolare: 247,09 g/mol
MDL Number: MFCD00185685
CAS Number: 89827-45-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR