Stampa…

2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine ≥98.0% (by total nitrogen basis)

Fornitore: TCI

B4342-1G B4342-5G
TCIAB4342-1GEA 79 EUR
TCIAB4342-1G TCIAB4342-5G
2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine ≥98.0% (by total nitrogen basis)
2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine
Formula: C₂₄H₂₈N₂O₄
MDL Number: MFCD00142492
CAS Number: 68842-66-0

Visualizza i codici

Risultati del test di specificazione

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 640.0 to 644.0 nm (CHCl3)
Absorbance (E1%1cm) min. 9100 (CHCl3, 640.0 to 644.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR