10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate) for chemiluminescence research
Fornitore: TCI
|
Attenzione
|
Synonyms:
MMT
B4339-100MG
B4339-1G
TCIAB4339-100MGEA
46
EUR
TCIAB4339-100MG
TCIAB4339-1G
10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate) for chemiluminescence research
10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate)
Formula:
C₄₆H₃₆N₂O₈ Peso molecolare: 744.80 g/mol |
CAS Number:
469865-01-8 |
Risultati del test di specificazione
Chemiluminescence test (Superoxide radical) | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...