Stampa…
trans-4-Bromocinnamaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: (E)-3-(4-Bromophenyl)acrylaldehyde
B4337-1G B4337-5G
TCIAB4337-1GEA 40 EUR
TCIAB4337-1G TCIAB4337-5G
trans-4-Bromocinnamaldehyde ≥98.0% (by GC)
trans-4-Bromocinnamaldehyde
Formula: C₉H₇BrO
Peso molecolare: 211,06 g/mol
MDL Number: MFCD03032509
CAS Number: 49678-04-8
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 85.0 deg-C