cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
cis-N1-Boc-1,2-cyclohexanediamine, cis-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, cis-2-Amino-1-(Boc-amino)cyclohexane, tert-Butyl (cis-2-aminocyclohexyl)carbamate
B4331-1G
B4331-5G
TCIAB4331-1GEA
118
EUR
TCIAB4331-1G
TCIAB4331-5G
cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥97.0% (by GC, titration analysis)
cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
Formula:
C₁₁H₂₂N₂O₂ Peso molecolare: 214,31 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD07367780 CAS Number: 184954-75-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...