4-Bromo-2-tert-butylphenol ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B4315-1G
B4315-5G
TCIAB4315-1GEA
84
EUR
TCIAB4315-1G
TCIAB4315-5G
4-Bromo-2-tert-butylphenol ≥95.0% (by GC)
4-Bromo-2-tert-butylphenol
Formula:
C₁₀H₁₃BrO Peso molecolare: 229,12 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00192645 CAS Number: 10323-39-4 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...