Stampa…

2-Bromo-5-iodo-3-methylpyridine ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromo-5-iodo-3-picoline

B4312-1G B4312-5G
TCIAB4312-1GEA 44 EUR
TCIAB4312-1G TCIAB4312-5G
2-Bromo-5-iodo-3-methylpyridine ≥96.0% (by GC)
2-Bromo-5-iodo-3-methylpyridine
Formula: C₆H₅BrIN
Peso molecolare: 297,92 g/mol
MDL Number: MFCD09037466
CAS Number: 65550-78-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR