5'-Bromo-m-terphenyl ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₈H₁₃Br |
MDL Number:
MFCD00196170 CAS Number: 103068-20-8 UN: 3152 ADR: 9,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...