Stampa…

N-Boc-D-glutamic acid 1-benzyl ester ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: 1-Benzyl N-Boc-D-glutamate, Boc-D-Glu-OBzl, N-Boc-D-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-D-glutamic Acid 1-Benzyl Ester

B4303-5G B4303-25G
TCIAB4303-5GEA 37 EUR
TCIAB4303-5G TCIAB4303-25G
N-Boc-D-glutamic acid 1-benzyl ester ≥98.0% (by HPLC, titration analysis)
N-Boc-D-glutamic acid 1-benzyl ester
Formula: C₁₇H₂₃NO₆
Peso molecolare: 337.37 g/mol
MDL Number: MFCD00038266
CAS Number: 34404-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16970d6c7bd454d6f6725e1dffe3480d4bbb0802&vid=e1c03f714a3f4896324f82b8fa1a6a04d2e9688a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C
Specific rotation [a]20/D +28.0 to +31.0 deg (C=0.7, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR