Stampa…

N-ɑ-Boc-L-tryptophanol ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: (S)-2-(tert-Butoxycarbonyl)amino-3-(3-indolyl)propanol, N-ɑ-(tert-Butoxycarbonyl)-L-tryptophanol, (S)-2-(Boc-amino)-3-(3-indolyl)propanol, Boc-Tryptophanol, N-Boc-L-tryptophanol

B4287-1G B4287-5G
TCIAB4287-1GEA 28 EUR
TCIAB4287-1G TCIAB4287-5G
N-ɑ-Boc-L-tryptophanol ≥98.0% (by HPLC, total nitrogen)
N-alpha-Boc-L-tryptophanol
Formula: C₁₆H₂₂N₂O₃
Peso molecolare: 290,36 g/mol
MDL Number: MFCD00235953
CAS Number: 82689-19-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 120.0 to 124.0 deg-C
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR