Stampa…

3-(Boc-amino)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

B4279-1G B4279-5G
TCIAB4279-1GEA 48 EUR
TCIAB4279-1G TCIAB4279-5G
3-(Boc-amino)phenylboronic acid (contains varying amounts of Anhydride)
3-(Boc-amino)phenylboronic acid
Formula: C₁₁H₁₆BNO₄
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR