9,9-Bis(4-glycidyloxyphenyl)fluorene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4,4'-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane), 2,2'-((((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(methylene))bis(oxirane)
B4269-5G
B4269-25G
TCIAB4269-5GEA
30
EUR
TCIAB4269-5G
TCIAB4269-25G
9,9-Bis(4-glycidyloxyphenyl)fluorene ≥98.0% (by GC)
9,9-Bis(4-glycidyloxyphenyl)fluorene
Formula:
C₃₁H₂₆O₄ Peso molecolare: 462,55 g/mol |
MDL Number:
MFCD02171148 CAS Number: 47758-37-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...