Stampa…

tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: [1-(Hydroxymethyl)cyclopropyl]carbamic acid tert-Butyl ester, [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol

B4251-1G B4251-200MG
TCIAB4251-1GEA 106 EUR
TCIAB4251-1G TCIAB4251-200MG
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate ≥97.0% (by GC)
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate
Formula: C₉H₁₇NO₃
Peso molecolare: 187,2362 g/mol
MDL Number: MFCD09749954
CAS Number: 107017-73-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR