Stampa…

4-Bromo-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

B4245-1G B4245-5G
TCIAB4245-1GEA 35 EUR
TCIAB4245-1G TCIAB4245-5G
4-Bromo-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Bromo-DL-phenylalanine
Formula: C₉H₁₀BrNO₂
Peso molecolare: 244.09 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002599
CAS Number: 14091-15-7
EINECS: 237-941-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR