Stampa…

4-Bromo-2-chlorobenzonitrile ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

B4241-1G B4241-5G
TCIAB4241-1GEA 16 EUR
TCIAB4241-1G TCIAB4241-5G
4-Bromo-2-chlorobenzonitrile ≥97.0% (by GC)
4-Bromo-2-chlorobenzonitrile
Formula: C₇H₃BrClN
Peso molecolare: 216,46 g/mol
MDL Number: MFCD00040883
CAS Number: 154607-01-9
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR