Stampa…

2,5-Bis[(trimethylsilyl)ethynyl]thiophene ≥96.0% (by GC)

Fornitore: TCI EUROPE

B4240-1G B4240-5G
TCIAB4240-1GEA 128 EUR
TCIAB4240-1G TCIAB4240-5G
2,5-Bis[(trimethylsilyl)ethynyl]thiophene ≥96.0% (by GC)
2,5-Bis[(trimethylsilyl)ethynyl]thiophene
Formula: C₁₄H₂₀SSi₂
Peso molecolare: 276,54 g/mol
MDL Number: MFCD18207392
CAS Number: 79109-69-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR