9-Bromo-9,10-dihydro-9,10-ethenoanthracene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₆H₁₁Br |
CAS Number:
126690-96-8 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...