Stampa…

tert-Butyl-2-amino-4-fluorophenylcarbamate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: tert-Butyl (2-Amino-4-fluorophenyl)carbamate, N1-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl 2-amino-4-fluorophenylcarbamate, (2-Amino-4-fluorophenyl)carbamic acid tert-Butyl ester, N1-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine

B4236-200MG B4236-1G
TCIAB4236-200MGEA 28 EUR
TCIAB4236-200MG TCIAB4236-1G
tert-Butyl-2-amino-4-fluorophenylcarbamate ≥98.0% (by titrimetric analysis)
tert-Butyl-2-amino-4-fluorophenylcarbamate
Formula: C₁₁H₁₅FN₂O₂
Peso molecolare: 226,2500 g/mol
MDL Number: MFCD09701246
CAS Number: 579474-47-8

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR