Stampa…

Bromobimane (mBBr) ≥98.0% (by HPLC)

Fornitore: TCI

B4220-20MG B4220-100MG
TCIAB4220-20MGEA 125 EUR
TCIAB4220-20MG TCIAB4220-100MG
Bromobimane (mBBr) ≥98.0% (by HPLC)
Bromobimane (mBBr)
Formula: C₁₀H₁₁BrN₂O₂
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00036951
CAS Number: 71418-44-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR