Butyraldehyde dibutyl acetal ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,1-Dibutoxybutane, Butyraldehyde Dibutyl Acetal
B4218-5G
B4218-25G
TCIAB4218-5GEA
134
EUR
TCIAB4218-5G
TCIAB4218-25G
Butyraldehyde dibutyl acetal ≥98.0% (by GC)
Butyraldehyde dibutyl acetal
Formula:
C₁₂H₂₆O₂ Peso molecolare: 202,34 g/mol Punto di ebollizione: 214°C |
MDL Number:
MFCD00561028 CAS Number: 5921-80-2 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...