4-Bromo-2-thiophenecarbonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
4-Bromothiophene-2-carbonitrile, 4-Bromo-2-cyanothiophene
Formula:
C₅H₂BrNS |
MDL Number:
MFCD00114951 CAS Number: 18791-99-6 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...