Stampa…

2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane ≥98.0% (by GC)

Fornitore: TCI EUROPE

B4168-1G B4168-5G
TCIAB4168-1GEA 96 EUR
TCIAB4168-1G TCIAB4168-5G
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane ≥98.0% (by GC)
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane
Formula: C₇BrF₁₅O₂
Peso molecolare: 480,96 g/mol
CAS Number: 1482416-44-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR