Stampa…

1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane ≥94.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane

B4167-1G B4167-5G
TCIAB4167-1GEA 85 EUR
TCIAB4167-1G TCIAB4167-5G
1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane ≥94.0% (by GC)
1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane
Formula: C₁₆BrF₃₃O₅
Peso molecolare: 979,02 g/mol
CAS Number: 1482416-52-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR