Stampa…

8-Bromoisoquinoline ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B4144-1G B4144-5G
TCIAB4144-1GEA 49 EUR
TCIAB4144-1G TCIAB4144-5G
8-Bromoisoquinoline ≥97.0% (by GC, titration analysis)
8-Bromoisoquinoline
Formula: C₉H₆BrN
Peso molecolare: 208,0600 g/mol
MDL Number: MFCD04973298
CAS Number: 63927-22-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR