Stampa…

N,N-Bis[2-(p-tolylsulphonyloxy)ethyl]-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (Tosylazanediyl)bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulphonamide

B4136-5G B4136-25G
TCIAB4136-5GEA 38 EUR
TCIAB4136-5G TCIAB4136-25G
N,N-Bis[2-(p-tolylsulphonyloxy)ethyl]-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N,N-Bis[2-(p-tolylsulphonyloxy)ethyl]-p-toluenesulphonamide
Formula: C₂₅H₂₉NO₈S₃
Peso molecolare: 567,69 g/mol
MDL Number: MFCD00026000
CAS Number: 16695-22-0
EINECS: 412-920-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR