Stampa…

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (R)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol, (R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol

B4131-5G B4131-25G
TCIAB4131-5GEA 31 EUR
TCIAB4131-5G TCIAB4131-25G
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Formula: C₁₀H₈F₆O
Peso molecolare: 258,16 g/mol
MDL Number: MFCD03093010
CAS Number: 127852-28-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C
Specific rotation [a]20/D +26.0 to +29.0 deg (C=1、CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR