Stampa…

5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98.0% (by HPLC) for biochemical research

Fornitore: TCI EUROPE

Synonyms: Magenta-GlcA CHA Salt, Magenta-Gluc CHA Salt, 5-Bromo-6-chloro-3-indolyl-β-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt

B4128-10MG B4128-100MG
TCIAB4128-10MGEA 22 EUR
TCIAB4128-10MG TCIAB4128-100MG
5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98.0% (by HPLC) for biochemical research
5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt
Formula: C₁₄H₁₃BrClNO₇
Peso molecolare: 521,79 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00153929
CAS Number: 144110-43-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.0 to 5.6 %
Specific rotation [a]20/D -66.0 to -70.0 deg (C=1,DMF (50vol%))
beta-Glucuronidase detection test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR