ω,3-Dibromoacetophenone ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Bromo-1-(3-bromophenyl)ethanone, 2-Bromo-1-(3-bromophenyl)-1-ethanone, α,3-Dibromoacetophenone, 2,3'-Dibromoacetophenone, 3-Bromophenacyl bromide
B4125-5G
B4125-25G
TCIAB4125-5GEA
80
EUR
TCIAB4125-5G
TCIAB4125-25G
ω,3-Dibromoacetophenone ≥98.0% (by titrimetric analysis)
ω,3-Dibromoacetophenone
Formula:
C₈H₆Br₂O Peso molecolare: 277,94 g/mol |
MDL Number:
MFCD00082698 CAS Number: 18523-22-3 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...