Stampa…

ω-Bromo-4-hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide

B4115-5G B4115-25G
TCIAB4115-5GEA 32 EUR
TCIAB4115-5G TCIAB4115-25G
ω-Bromo-4-hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
ω-Bromo-4-hydroxyacetophenone
Formula: C₈H₇BrO₂
Peso molecolare: 215,05 g/mol
MDL Number: MFCD00072424
CAS Number: 2491-38-5
EINECS: 219-655-0
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR