Stampa…

9-(2-Bromophenyl)-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4109-1G B4109-5G
TCIAB4109-1GEA 23 EUR
TCIAB4109-1G TCIAB4109-5G
9-(2-Bromophenyl)-9H-carbazole ≥98.0% (by GC)
9-(2-Bromophenyl)-9H-carbazole
Formula: C₁₈H₁₂BrN
Peso molecolare: 322,21 g/mol
MDL Number: MFCD23135883
CAS Number: 902518-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e480ce788979bb046395d7b061aa16f0a282cb&vid=79e480ce788979bb046395d7b061aa16f0a282cb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR