Stampa…

Benserazide Hydrochloride ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Benserazide HCI, DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride, 2-Amino-3-hydroxy-N'-(2,3,4-trihydroxybenzyl)propanehydrazide hydrochloride

B4108-1G B4108-5G
TCIAB4108-1GEA 53 EUR
TCIAB4108-1G TCIAB4108-5G
Benserazide Hydrochloride ≥97.0% (by HPLC, titration analysis)
Benserazide hydrochloride
Formula: C₁₀H₁₅N₃O₅
Peso molecolare: 293.70 g/mol
MDL Number: MFCD00078571
CAS Number: 14919-77-8
EINECS: 238-991-9
Merck Index: 14,01050

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR