Stampa…

(±)-2-Bromopropionyl chloride ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Bromopropionylchloride

B4107-25G B4107-100G
TCIAB4107-25GEA 60 EUR
TCIAB4107-25G TCIAB4107-100G
(±)-2-Bromopropionyl chloride ≥97.0% (by titrimetric analysis)
(±)-2-Bromopropionyl chloride
Formula: C₃H₄BrClO
Peso molecolare: 171,42 g/mol
Punto di ebollizione: 131°C
Flash Pt: 51°C
MDL Number: MFCD00000711
CAS Number: 7148-74-5
EINECS: 230-466-2
UN: 2920
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR