Stampa…

Bis[(pinacolato)boryl]methane ≥95.0% (by GC)

Fornitore: TCI

Synonyms: 2,2'-Methylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methane

B4103-1G B4103-5G
TCIAB4103-1GEA 72 EUR
TCIAB4103-1G TCIAB4103-5G
Bis[(pinacolato)boryl]methane ≥95.0% (by GC)
Bis[(pinacolato)boryl]methane
Formula: C₁₃H₂₆B₂O₄
Peso molecolare: 267.97 g/mol
MDL Number: MFCD27977747
CAS Number: 78782-17-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR