6-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromo-6-methylbenzoic acid
B4097-1G
B4097-5G
TCIAB4097-1GEA
52
EUR
TCIAB4097-1G
TCIAB4097-5G
6-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
6-Bromo-o-toluic acid
Formula:
C₈H₇BrO₂ Peso molecolare: 215,05 g/mol |
MDL Number:
MFCD01310788 CAS Number: 90259-31-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...