Stampa…

9,9'-Bianthracene ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 9,9'-Bianthracenyl, 9,9'-Bianthryl

B4095-1G B4095-5G
TCIAB4095-1GEA 151 EUR
TCIAB4095-1G TCIAB4095-5G
9,9'-Bianthracene ≥98.0% (by HPLC)
9,9'-Bianthracene
Formula: C₂₈H₁₈
Peso molecolare: 354,45 g/mol
Punto di fusione: 306°C
MDL Number: MFCD00130216
CAS Number: 1055-23-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR