2-(4-tert-Butylphenyl)pyridine ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(4-(tert-Butyl)phenyl)pyridine
B4092-200MG
B4092-1G
TCIAB4092-200MGEA
96
EUR
TCIAB4092-200MG
TCIAB4092-1G
2-(4-tert-Butylphenyl)pyridine ≥97.0% (by GC)
2-(4-tert-Butylphenyl)pyridine
Formula:
C₁₅H₁₇N Peso molecolare: 211,31 g/mol |
MDL Number:
MFCD07775634 CAS Number: 524713-66-4 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...