Stampa…

4-Bromo-2-methoxyphenol ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromoguaiacol, 5-Brom-2-hydroxyanisole

B4056-5G B4056-25G
TCIAB4056-5GEA 67 EUR
TCIAB4056-5G TCIAB4056-25G
4-Bromo-2-methoxyphenol ≥97.0% (by GC, titration analysis)
4-Bromo-2-methoxyphenol
Formula: C₇H₇BrO₂
Peso molecolare: 203,04 g/mol
Punto di ebollizione: 133°C
MDL Number: MFCD00051937
CAS Number: 7368-78-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 33.0 to 37.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR