Stampa…

4-(4-Bromophenylazo)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromo-4'-hydroxyazobenzene

B4053-200MG B4053-1G
TCIAB4053-200MGEA 116 EUR
TCIAB4053-200MG TCIAB4053-1G
4-(4-Bromophenylazo)phenol ≥98.0% (by GC)
4-(4-Bromophenylazo)phenol
Formula: C₁₂H₉BrN₂O
Peso molecolare: 277,12 g/mol
MDL Number: MFCD00093929
CAS Number: 3035-94-7

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR