Stampa…

(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate, tert-Butyl L-Pyroglutamate, tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, tert-Butyl (S)-2-pyrrolidone-5-carboxylate

B4026-1G B4026-5G
TCIAB4026-1GEA 16 EUR
TCIAB4026-1G TCIAB4026-5G
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥98.0% (by GC)
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
Formula: C₉H₁₅NO₃
Peso molecolare: 185,22 g/mol
MDL Number: MFCD06659481
CAS Number: 35418-16-7
EINECS: 252-555-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 108.0 deg-C
Specific rotation [a]20/D +8.0 to +10.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR