1-Bromo-4-(phenylethynyl)benzene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₉Br |
MDL Number:
MFCD01596910 CAS Number: 13667-12-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...