Stampa…

4-Bromo-3-fluorobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4022-5G B4022-25G
TCIAB4022-5GEA 60 EUR
TCIAB4022-5G TCIAB4022-25G
4-Bromo-3-fluorobenzonitrile ≥98.0% (by GC)
4-Bromo-3-fluorobenzonitrile
Formula: C₇H₃BrFN
Peso molecolare: 200,01 g/mol
MDL Number: MFCD07782069
CAS Number: 133059-44-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR