Stampa…

4-Bromo-2,6-difluorobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

B4021-1G B4021-5G
TCIAB4021-1GEA 44 EUR
TCIAB4021-1G TCIAB4021-5G
4-Bromo-2,6-difluorobenzonitrile ≥98.0% (by GC)
4-Bromo-2,6-difluorobenzonitrile
Formula: C₇H₂BrF₂N
Peso molecolare: 218 g/mol
MDL Number: MFCD01310981
CAS Number: 123843-67-4
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR