2-Bromo-2',4'-difluoroacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Bromo-1-(2,4-difluorophenyl)ethanone, 2,4-Difluorophenacyl bromide
B4020-5G
B4020-25G
TCIAB4020-5GEA
61
EUR
TCIAB4020-5G
TCIAB4020-25G
2-Bromo-2',4'-difluoroacetophenone ≥98.0% (by GC)
2-Bromo-2',4'-difluoroacetophenone
Formula:
C₈H₅BrF₂O Peso molecolare: 235,03 g/mol Flash Pt: 51°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00142822 CAS Number: 102429-07-2 UN: 2925 ADR: 4.1,II |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow to Light orange powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 33.0 to 37.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...