Stampa…
2-Bromo-6-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Attenzione
B4017-1G B4017-5G
TCIAB4017-1GEA 27 EUR
TCIAB4017-1G TCIAB4017-5G
2-Bromo-6-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-6-fluorobenzoic acid
Formula: C₇H₄BrFO₂
Peso molecolare: 219.01 g/mol
MDL Number: MFCD01569539
CAS Number: 2252-37-1
Visualizza i codici
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 158.0 deg-C