(±)-Boc-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate
B4011-1G
B4011-5G
TCIAB4011-1GEA
34
EUR
TCIAB4011-1G
TCIAB4011-5G
(±)-Boc-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)
(±)-Boc-α-phosphonoglycine trimethyl ester
Formula:
C₁₀H₂₀NO₇P Peso molecolare: 297,24 g/mol |
MDL Number:
MFCD02178893 CAS Number: 89524-98-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...